Assay Details
| Gene Symbol: P2ry10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172435.3 | NP_766023.1 | 3 | 2665 | 134 | |
| GenBank mRNA | AK020001.1 | - | 3 | 2544 | 134 | |
| AK088234.1 | - | 3 | 2666 | 134 | ||
| BC053707.1 | - | 3 | 2496 | 30072257 | 134 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105963 | AK088234.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105963 | BC053707.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105963 | NM_172435.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2105963 | AK020001.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 679963 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 516264 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 708786 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 948637 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 335099 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 396994 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 284934 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 145128 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 88519 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441847 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 103361 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140025 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103504 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105963 | AK088234.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105963 | BC053707.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105963 | NM_172435.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2105963 | AK020001.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P301367 | AK088234.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P301367 | BC053707.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P301367 | AK038124.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P301367 | NM_172435.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P301367 | AK020001.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586761 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586740 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586730 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586736 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586743 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586759 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586747 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586728 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586744 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586735 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586745 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586741 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586726 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586733 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32586725 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140025 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P103504 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09386761 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09386746 | - | - |
| Whole Mouse Genome G4122A | A_51_P301367 | AK088234.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301367 | BC053707.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301367 | AK038124.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301367 | NM_172435.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301367 | AK020001.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129262 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140025 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P103504 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09386761 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14992921 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15086240 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14992924 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15086230 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15086245 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14992919 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14992923 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15086231 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15086236 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15086246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32586821 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586779 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586749 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586728 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586736 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586785 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586793 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586796 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586757 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586808 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586784 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586747 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586755 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586815 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586730 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586787 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586761 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14558064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586782 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14954330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586797 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586726 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586772 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586770 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586802 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586816 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586746 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586740 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586743 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586734 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586733 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586759 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586819 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586771 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586820 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586725 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586812 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586783 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14558071 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586739 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586731 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586780 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14581278 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586814 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586790 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14906647 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586778 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586774 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586805 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586723 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586786 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586818 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586766 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586735 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32586744 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10601419 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601418 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601420 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452815_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10601416 | - | - |
| Mouse Genome 430 2.0 Array | 1452815_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452815_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78826_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78826_at | - | - |
| Mouse Expression Set 430 | 1452815_at | - | - |
| Mouse Expression Set 430 | 1452815_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452815_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452815_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 563740 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6370039 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6370039 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6840204 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006370039 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006840204 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1224813 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1224483 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1224483 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1224813 | - | - |
| mouse-6 v1.1 (Illumina) | scl0002943.1_105-S | - | - |
| mouse-6 v1.1 (Illumina) | scl54689.4_655-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6840204 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6370039 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002943.1_105-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl54689.4_655-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6370039 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6840204 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2954781 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1224483 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2723040 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1224483 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2723040 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2954781 | - | - |

P2ry10
Genomic Map