Assay Details
| Gene Symbol: Gpr62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159652.1 | NP_001153124.1 | 1 | 1056 | 59 | |
| GenBank mRNA | AK005968.1 | - | 1 | 47 | 59 | |
| AK015060.1 | - | 1 | 34 | 59 | ||
| AK015270.1 | - | 1 | 43 | 59 | ||
| AK016613.1 | - | 1 | 34 | 59 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gpr62
Genomic Map