Assay Details
| Gene Symbol: Ffar3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033316.2 | NP_001028488.1 | 2 | 745 | 140 | |
| GenBank mRNA | AK148616.1 | - | 2 | 746 | 140 | |
| BC109330.2 | - | 1 | 958 | 40051470 | 140 | |
| BC125009.1 | - | 1 | 958 | 40051462 | 140 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
immune system process
mucosal immune response
positive regulation of cytokine production involved in immune response
positive regulation of acute inflammatory response to non-antigenic stimulus
regulation of heart rate by chemical signal
inflammatory response
signal transduction
G-protein coupled receptor signaling pathway
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
regulation of norepinephrine secretion
positive regulation of chemokine production
glucose homeostasis
positive regulation of action potential
negative regulation of blood pressure
regulation of hormone biosynthetic process
cellular response to fatty acid
regulation of peptide hormone secretion
positive regulation of glucose import in response to insulin stimulus
signal transducer activity
G-protein coupled receptor activity
lipid binding
immune system process
mucosal immune response
positive regulation of cytokine production involved in immune response
positive regulation of acute inflammatory response to non-antigenic stimulus
regulation of heart rate by chemical signal
inflammatory response
signal transduction
G-protein coupled receptor signaling pathway
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
regulation of norepinephrine secretion
positive regulation of chemokine production
glucose homeostasis
positive regulation of action potential
negative regulation of blood pressure
regulation of hormone biosynthetic process
cellular response to fatty acid
regulation of peptide hormone secretion
positive regulation of glucose import in response to insulin stimulus
signal transducer activity
G-protein coupled receptor activity
lipid binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P106699 | NM_001033316.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P106699 | AK148616.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P106699 | BC125009.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 4112 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 103606 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P106699 | NM_001033316.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P106699 | AK148616.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P106699 | BC125009.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033728 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033735 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033725 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033748 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033743 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033731 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033720 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033726 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033751 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033742 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033733 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033717 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033744 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033736 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033745 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033730 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033716 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25033755 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06155790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25033717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06436079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06336265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06436075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06436081 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06486372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06573313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033712 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033746 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06598663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033731 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033726 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06436084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06436077 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25033751 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10562127 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562129 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562128 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562126 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10562125 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700024792.mm.1 | NM_001033316.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700024792.mm.1 | NM_001033316.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700024792.mm.1 | TC0700002777.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700024793.mm.1 | NM_001033316.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700024793.mm.1 | NM_001033316.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700024793.mm.1 | TC0700002777.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700024795.mm.1 | NM_001033316.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700024795.mm.1 | NM_001033316.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700024795.mm.1 | TC0700002777.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700024794.mm.1 | TC0700002777.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 769770 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1236703 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2869075 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2869075 | - | - |

Ffar3
Genomic Map