Assay Details
| Gene Symbol: Flg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013804.1 | NP_001013826.1 | 3766 | 154 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P1035965 | NM_001013804.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P1035965 | AK036878.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P1035965 | NM_001013804.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P1035965 | AK036878.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 900390 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 928858 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 817811 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 542817 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 163671 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158188 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P1035965 | NM_001013804.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P1035965 | AK036878.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P1035965 | NM_001013804.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P1035965 | AK036878.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00700395 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04990804 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158188 | - | - |
| Whole Mouse Genome G4122A | A_52_P1035965 | NM_001013804.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1035965 | AK036878.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1035965 | NM_001013804.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1035965 | AK036878.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04990804 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22335074 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02700170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02700187 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03095645 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335009 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335014 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335019 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335029 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335083 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335064 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335025 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03043417 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335030 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335059 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02700184 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335053 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335054 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335008 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335028 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335050 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335040 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335007 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335017 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02700181 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335073 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03095646 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02700166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335038 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02700180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335074 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335067 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02332689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335005 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02700176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02700156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02700173 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335049 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335068 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335056 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02945810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02700186 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03095642 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335016 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02700179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335036 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335001 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03043416 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02700160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22335034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02700174 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10493919 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493920 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493917 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493918 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10493916 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300006471.mm.1 | NM_001013804.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006471.mm.1 | NM_001013804.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006471.mm.1 | TC0300000843.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006480.mm.1 | NM_001013804.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006480.mm.1 | NM_001013804.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006480.mm.1 | TC0300000843.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006478.mm.1 | NM_001013804.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006478.mm.1 | TC0300000843.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006473.mm.1 | NM_001013804.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006473.mm.1 | NM_001013804.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006473.mm.1 | TC0300000843.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006479.mm.1 | NM_001013804.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006479.mm.1 | NM_001013804.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006479.mm.1 | TC0300000843.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006472.mm.1 | NM_001013804.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006472.mm.1 | NM_001013804.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006472.mm.1 | TC0300000843.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006474.mm.1 | NM_001013804.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006474.mm.1 | NM_001013804.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006474.mm.1 | TC0300000843.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2528661 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1257656 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2826699 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2826699 | - | - |

Flg2
Genomic Map