Assay Details
| Gene Symbol: Gm6498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003630.2 | 2 - 3 | 520 | 80 | ||
| GenBank mRNA | AK033118.1 | - | 2 - 3 | 520 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | pellino 2 | Peli2 |
AW047589, BB129927 |
Sense | Chr. 14 - 48120778 - 48308103 on NCBI Build GRCm38 | |
| Downstream | transmembrane protein 260 | Tmem260 |
5930435P15, 6720456H20Rik |
217.7kb | Sense | Chr. 14 - 48446128 - 48520708 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK033118.1 transcripts.
Set Membership: |
|

Gm6498
Genomic Map