| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700003222.mm.1 |
NR_073028.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003222.mm.1 |
TC0700000414.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003221.mm.1 |
NR_073028.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003221.mm.1 |
TC0700000414.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003229.mm.1 |
NR_073028.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003229.mm.1 |
TC0700000414.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003225.mm.1 |
NR_073028.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003225.mm.1 |
TC0700000414.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003230.mm.1 |
NR_073028.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003230.mm.1 |
TC0700000414.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003223.mm.1 |
NR_073028.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003223.mm.1 |
TC0700000414.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003227.mm.1 |
NR_073028.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003227.mm.1 |
TC0700000414.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003228.mm.1 |
NR_073028.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003228.mm.1 |
TC0700000414.mm.1 |
MTA TC |