Assay Details
| Gene Symbol: Krt40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039666.1 | NP_001034755.1 | 3 - 4 | 691 | 93 | |
| GenBank mRNA | BC107359.1 | - | 3 - 4 | 691 | 93 | |
| BC107360.2 | - | 3 - 4 | 689 | 93 | ||
| BC119522.2 | - | 3 - 4 | 691 | 40055959 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Krt40
Genomic Map