Assay Details
| Gene Symbol: Fmod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021355.3 | NP_067330.1 | 2 | 814 | 115 | |
| GenBank mRNA | AK032302.1 | - | 2 | 815 | 115 | |
| AK054183.1 | - | 2 | 871 | 115 | ||
| AK080601.1 | - | 2 | 832 | 115 | ||
| AK082593.1 | - | 2 | 831 | 115 | ||
| BC052673.1 | - | 2 | 823 | 115 | ||
| BC064779.1 | - | 2 | 787 | 30058603 | 115 | |
| X94998.1 | - | 2 | 786 | 115 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fmod
Genomic Map