Assay Details
| Gene Symbol: Lef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276403.1 | NP_001263332.1 | 11 - 12 | 1022 | 75 | |
| NM_010703.4 | NP_034833.2 | 11 - 12 | 2374 | 75 | ||
| GenBank mRNA | AK083328.1 | - | 11 - 12 | 1017 | 75 | |
| AK158769.1 | - | 11 - 12 | 2356 | 75 | ||
| X58636.1 | - | 11 - 12 | 2188 | 75 | ||
More Information
Set Membership: |
|

Lef1
Genomic Map