Assay Details
| Gene Symbol: Lef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276402.1 | NP_001263331.1 | 3 - 4 | 1584 | 86 | |
| NM_001276403.1 | NP_001263332.1 | 3 - 4 | 232 | 86 | ||
| NM_010703.4 | NP_034833.2 | 3 - 4 | 1584 | 86 | ||
| GenBank mRNA | AK028246.1 | - | 3 - 4 | 1206 | 86 | |
| AK034819.1 | - | 3 - 4 | 1057 | 86 | ||
| AK083328.1 | - | 3 - 4 | 227 | 86 | ||
| AK131979.1 | - | 3 - 4 | 1566 | 86 | ||
| AK158769.1 | - | 3 - 4 | 1566 | 86 | ||
| AK169530.1 | - | 2 - 3 | 218 | 86 | ||
| BC038305.1 | - | 3 - 4 | 367 | 86 | ||
| BC057543.1 | - | 3 - 4 | 1590 | 6401514 | 86 | |
| X58636.1 | - | 3 - 4 | 1398 | 86 | ||
More Information
Set Membership: |
|

Lef1
Genomic Map