Assay Details
| Gene Symbol: Lef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276402.1 | NP_001263331.1 | 8 - 9 | 2199 | 97 | |
| NM_001276403.1 | NP_001263332.1 | 9 - 10 | 931 | 97 | ||
| NM_010703.4 | NP_034833.2 | 9 - 10 | 2283 | 97 | ||
| GenBank mRNA | AK028246.1 | - | 8 - 9 | 1821 | 97 | |
| AK034819.1 | - | 8 - 9 | 1672 | 97 | ||
| AK083328.1 | - | 9 - 10 | 926 | 97 | ||
| AK131979.1 | - | 8 - 9 | 2180 | 97 | ||
| AK158769.1 | - | 9 - 10 | 2265 | 97 | ||
| BC057543.1 | - | 8 - 9 | 2205 | 6401514 | 97 | |
| X58636.1 | - | 9 - 10 | 2097 | 97 | ||
More Information
Set Membership: |
|

Lef1
Genomic Map