Assay Details
| Gene Symbol: Egr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018781.3 | NP_061251.1 | 1 - 2 | 151 | 92 | |
| GenBank mRNA | AB221656.1 | - | 1 - 2 | 151 | 92 | |
| AF132128.1 | - | 1 - 2 | 151 | 92 | ||
| BC100734.1 | - | 1 - 2 | 150 | 92 | ||
| BC103569.1 | - | 1 - 2 | 146 | 92 | ||
More Information
Set Membership: |
|

Egr3
Genomic Map