Assay Details
| Gene Symbol: Gsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206367.1 | NP_001193296.1 | 13 - 14 | 1791 | 88 | |
| NM_001206368.1 | NP_001193297.1 | 12 - 13 | 1707 | 88 | ||
| NM_001206369.1 | NP_001193298.1 | 12 - 13 | 1752 | 88 | ||
| NM_146120.4 | NP_666232.2 | 12 - 13 | 1814 | 88 | ||
| GenBank mRNA | AK076156.1 | - | 12 - 13 | 1754 | 88 | |
| AK089934.1 | - | 13 - 14 | 1746 | 88 | ||
| AK135632.1 | - | 12 - 13 | 1527 | 88 | ||
| AK143664.1 | - | 12 - 13 | 1781 | 88 | ||
| AK146969.1 | - | 13 - 14 | 1708 | 88 | ||
| AK148957.1 | - | 13 - 14 | 1771 | 88 | ||
| AK150359.1 | - | 13 - 14 | 1714 | 88 | ||
| AK151684.1 | - | 13 - 14 | 1733 | 88 | ||
| AK153029.1 | - | 13 - 14 | 1711 | 88 | ||
| AK166831.1 | - | 12 - 13 | 1727 | 88 | ||
| AK166932.1 | - | 13 - 14 | 1705 | 88 | ||
| AK167918.1 | - | 13 - 14 | 1721 | 88 | ||
| AK168461.1 | - | 13 - 14 | 1724 | 88 | ||
| AK168532.1 | - | 13 - 14 | 1725 | 88 | ||
| AK168707.1 | - | 12 - 13 | 1660 | 88 | ||
| AK168725.1 | - | 13 - 14 | 1708 | 88 | ||
| AK168829.1 | - | 12 - 13 | 1760 | 88 | ||
| AK169041.1 | - | 13 - 14 | 1711 | 88 | ||
| AK169670.1 | - | 13 - 14 | 1736 | 88 | ||
| BC023143.1 | - | 12 - 13 | 1752 | 3711602 | 88 | |
| BC060377.1 | - | 13 - 14 | 1739 | 4983664 | 88 | |
| J04953.1 | - | 11 - 12 | 1601 | 88 | ||
More Information
Set Membership: |
|

Gsn
Genomic Map