Assay Details
| Gene Symbol: Gsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206367.1 | NP_001193296.1 | 9 - 10 | 1185 | 114 | |
| NM_001206368.1 | NP_001193297.1 | 8 - 9 | 1101 | 114 | ||
| NM_001206369.1 | NP_001193298.1 | 8 - 9 | 1146 | 114 | ||
| NM_146120.4 | NP_666232.2 | 8 - 9 | 1208 | 114 | ||
| GenBank mRNA | AK076156.1 | - | 8 - 9 | 1148 | 114 | |
| AK089934.1 | - | 9 - 10 | 1140 | 114 | ||
| AK135632.1 | - | 7 - 8 | 921 | 114 | ||
| AK143664.1 | - | 8 - 9 | 1175 | 114 | ||
| AK146969.1 | - | 9 - 10 | 1102 | 114 | ||
| AK148957.1 | - | 9 - 10 | 1165 | 114 | ||
| AK150359.1 | - | 9 - 10 | 1108 | 114 | ||
| AK151684.1 | - | 9 - 10 | 1127 | 114 | ||
| AK153029.1 | - | 9 - 10 | 1105 | 114 | ||
| AK166831.1 | - | 8 - 9 | 1121 | 114 | ||
| AK166932.1 | - | 9 - 10 | 1099 | 114 | ||
| AK167918.1 | - | 9 - 10 | 1115 | 114 | ||
| AK168461.1 | - | 9 - 10 | 1118 | 114 | ||
| AK168532.1 | - | 9 - 10 | 1119 | 114 | ||
| AK168707.1 | - | 8 - 9 | 1054 | 114 | ||
| AK168725.1 | - | 9 - 10 | 1102 | 114 | ||
| AK168829.1 | - | 8 - 9 | 1153 | 114 | ||
| AK169041.1 | - | 9 - 10 | 1105 | 114 | ||
| AK169670.1 | - | 9 - 10 | 1130 | 114 | ||
| BC023143.1 | - | 8 - 9 | 1146 | 3711602 | 114 | |
| BC060377.1 | - | 9 - 10 | 1133 | 4983664 | 114 | |
| J04953.1 | - | 7 - 8 | 995 | 114 | ||
More Information
Set Membership: |
|

Gsn
Genomic Map