Assay Details
| Gene Symbol: Prpmp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024705.2 | NP_001019876.2 | 3 | 946 | 126 | |
| GenBank mRNA | BC055856.1 | - | 3 | 937 | 4910526 | 126 |
| BC061494.1 | - | 3 | 937 | 126 | ||
| BC080748.1 | - | 3 | 937 | 126 | ||
| BC092075.1 | - | 3 | 941 | 4920522 | 126 | |
| BC110664.1 | - | 3 | 944 | 4191067 | 126 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2050353 | NM_198669.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2050353 | BC011176.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2050353 | BC080748.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2050353 | BC092075.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2050353 | BC110664.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281824 | BC080748.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281824 | BC092075.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281824 | BC110664.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281824 | X63004.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281824 | NM_001024705.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281824 | BC080748.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281824 | BC092075.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281824 | BC110664.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281824 | X63004.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281824 | NM_001024705.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2050354 | BC080748.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2050354 | BC092075.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2050354 | BC110664.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2050354 | NM_001024705.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2050354 | BC055856.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2050354 | BC080748.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2050354 | BC092075.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2050354 | BC110664.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2050354 | NM_001024705.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2050354 | BC055856.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281824 | BC080748.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281824 | BC092075.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281824 | BC110664.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281824 | X63004.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281824 | NM_001024705.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281824 | BC080748.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281824 | BC092075.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281824 | BC110664.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281824 | X63004.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281824 | NM_001024705.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2050353 | NM_198669.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2050353 | BC011176.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2050353 | BC080748.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2050353 | BC092075.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2050353 | BC110664.1 | mouse_transcriptome |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24853176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24853195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05329780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24853182 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10548634 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548631 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548624 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548654 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548636 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548641 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548650 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548626 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548635 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548645 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548643 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548652 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548633 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548659 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548632 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548651 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548640 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548655 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548628 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548625 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421490_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10548653 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10548639 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10548656 | - | - |
| Mouse Genome 430 2.0 Array | 1421490_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1427745_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1427745_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1421490_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 381832_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 381832_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600025800.mm.1 | NM_001024705.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025800.mm.1 | NM_001024705.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025800.mm.1 | TC0600003281.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025802.mm.1 | NM_001024705.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025802.mm.1 | NM_001024705.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025802.mm.1 | TC0600003281.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025799.mm.1 | NM_001024705.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025799.mm.1 | NM_001024705.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025799.mm.1 | TC0600003281.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025801.mm.1 | NM_001024705.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025801.mm.1 | NM_001024705.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025801.mm.1 | TC0600003281.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427745_x_at | - | - |
| Mouse Expression Set 430 | 1427745_PM_x_at | - | - |
| Mouse Expression Set 430 | 1421490_at | - | - |
| Mouse Expression Set 430 | 1421490_PM_at | - | - |
| Murine Genome U74v2 Set | 98383_PM_r_at | - | - |
| Murine Genome U74v2 Set | 98382_PM_f_at | - | - |
| Murine Genome U74v2 Set | 98383_r_at | - | - |
| Murine Genome U74v2 Set | 98382_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421490_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421490_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427745_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427745_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421490_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421490_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 704565 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1242192 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2850496 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2850496 | - | - |

Prpmp5
Genomic Map