Assay Details
| Gene Symbol: Ppia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008907.1 | NP_032933.1 | 2 | 123 | 145 | |
| GenBank mRNA | AK002236.1 | - | 2 | 126 | 145 | |
| AK007981.1 | - | 2 | 128 | 145 | ||
| AK010649.1 | - | 2 | 126 | 145 | ||
| AK012531.1 | - | 2 | 126 | 145 | ||
| AK012541.1 | - | 2 | 122 | 145 | ||
| AK012663.1 | - | 2 | 126 | 145 | ||
| AK028210.1 | - | 2 | 126 | 145 | ||
| AK131942.1 | - | 2 | 126 | 145 | ||
| AK136788.1 | - | 2 | 126 | 145 | ||
| AK151347.1 | - | 2 | 126 | 145 | ||
| AK151583.1 | - | 2 | 215 | 145 | ||
| AK153030.1 | - | 2 | 125 | 145 | ||
| AK160631.1 | - | 2 | 126 | 145 | ||
| AK160780.1 | - | 2 | 154 | 145 | ||
| AK168035.1 | - | 2 | 126 | 145 | ||
| AK169813.1 | - | 2 | 126 | 145 | ||
| BC013713.1 | - | 2 | 114 | 145 | ||
| BC083076.1 | - | 2 | 114 | 6442005 | 145 | |
| BC087928.1 | - | 2 | 125 | 6533710 | 145 | |
| BC094272.1 | - | 2 | 114 | 2645114 | 145 | |
| BC094632.1 | - | 2 | 106 | 5055863 | 145 | |
| BC099478.1 | - | 2 | 117 | 30843162 | 145 | |
| DQ985731.1 | - | 2 | 82 | 145 | ||
| X52803.1 | - | 2 | 123 | 145 | ||
| Gene Symbol: E030024N20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033228.1 | 1 | 145 | |||
| GenBank mRNA | AK053172.1 | - | 1 | 145 | ||
| AK142495.1 | - | 1 | 145 | |||
| Gene Symbol: Gm17494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK138382.1 | - | 2 | 145 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ppia
Genomic Map