Assay Details
| Gene Symbol: Rgma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177740.5 | NP_808408.2 | 3 - 4 | 1002 | 122 | |
| GenBank mRNA | AJ557513.1 | - | 2 - 3 | 756 | 122 | |
| AK082560.1 | - | 2 - 3 | 899 | 122 | ||
| BC023870.1 | - | 2 - 3 | 810 | 5353958 | 122 | |
| BC059072.1 | - | 3 - 4 | 1002 | 6826181 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rgma
Genomic Map