| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100014285.mm.1 |
NM_177099.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100014285.mm.1 |
TC0100001721.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100014286.mm.1 |
NM_177099.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100014286.mm.1 |
TC0100001721.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100014287.mm.1 |
NM_177099.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100014287.mm.1 |
TC0100001721.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100014305.mm.1 |
NM_010094.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100014305.mm.1 |
TC0100001723.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100014306.mm.1 |
NM_010094.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100014306.mm.1 |
TC0100001723.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100014307.mm.1 |
NM_010094.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100014307.mm.1 |
TC0100001723.mm.1 |
MTA TC |