Assay Details
| Gene Symbol: Hspa1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010478.2 | NP_034608.2 | 1 | 2639 | 141 | |
| GenBank mRNA | AK159139.1 | - | 2607 | 141 | ||
| AK165042.1 | - | 1 | 2646 | 141 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156686 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156690 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156689 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156687 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156691 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 270343 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155999 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P304902 | AK078618.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P304902 | BC157921.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P304902 | AK171820.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P304902 | BC032044.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P304902 | BC054782.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P52803 | BC157921.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P52803 | M12571.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P52803 | M12572.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P52803 | BC054782.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P52803 | AK132331.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156800 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156687 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156691 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156689 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156686 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156677 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156745 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156697 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156744 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156752 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156780 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156710 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156667 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13101693 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13587175 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156767 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156777 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156747 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156670 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156713 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156766 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156778 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13564407 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13070479 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156741 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13460196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156673 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13079669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156677 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156708 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13564406 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156772 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156666 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156749 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156785 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156748 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156787 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156762 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156784 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156715 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156792 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156716 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13322045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13547681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156754 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13079673 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13295068 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13060484 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156703 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13403353 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156668 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13564410 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156782 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13564404 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13220993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13560606 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156691 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156760 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156694 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156663 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156793 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13485290 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10444590 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450371 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450368 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452318_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427127_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427126_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10444589 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10450367 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10450369 | - | - |
| Mouse Genome 430 2.0 Array | 1427127_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1427126_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1427127_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1427126_at | - | - |
| Mouse Genome 430 2.0 Array | 1452318_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1452318_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15511_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15511_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700018333.mm.1 | NM_010478.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018333.mm.1 | NM_010478.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018333.mm.1 | TC1700001935.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700018337.mm.1 | NM_010478.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018337.mm.1 | NM_010478.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018337.mm.1 | TC1700001935.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700018331.mm.1 | NM_010478.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018331.mm.1 | NM_010478.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018331.mm.1 | TC1700001935.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700018335.mm.1 | NM_010478.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018335.mm.1 | NM_010478.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700018335.mm.1 | TC1700001935.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452318_PM_a_at | - | - |
| Mouse Expression Set 430 | 1427127_PM_x_at | - | - |
| Mouse Expression Set 430 | 1452318_a_at | - | - |
| Mouse Expression Set 430 | 1427127_x_at | - | - |
| Mouse Expression Set 430 | 1427126_PM_at | - | - |
| Mouse Expression Set 430 | 1427126_at | - | - |
| Murine Genome U74v2 Set | 100946_PM_at | - | - |
| Murine Genome U74v2 Set | 100946_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452318_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452318_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427126_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427126_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427127_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427127_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427126_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452318_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427127_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427127_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427126_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452318_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 880965 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 105700270 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2498263 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2530672 | - | - |

Hspa1b
Genomic Map