Assay Details
| Gene Symbol: Proca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045516.2 | NP_001038981.2 | 3 - 4 | 596 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2110567 | AK160258.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2110567 | BY648103.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2110567 | NM_001045516.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 751590 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 529352 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 92633 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 152924 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 524286 | AK147447.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 524286 | BC145861.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 524286 | NM_009688.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 524286 | U36842.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 524286 | U88990.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 772477 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 743431 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 566618 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 656015 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 4526 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2110567 | AK160258.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2110567 | BY648103.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2110567 | NM_001045516.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2025123 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07407530 | - | - |
| Whole Mouse Genome G4122A | A_52_P453469 | AK160258.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P453469 | NM_001045516.2 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27941888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09110214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09246575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941884 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941893 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09520932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941892 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941904 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941903 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941899 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941897 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941922 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09104227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08986678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09513498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09110227 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09193086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09627679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941878 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08986672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27941881 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10379066 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379064 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379067 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10379063 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1442508_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10379060 | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 216974_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 216974_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100010576.mm.1 | NM_001045516.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010576.mm.1 | NM_001045516.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010576.mm.1 | TC1100001150.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100010583.mm.1 | NM_001045516.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010583.mm.1 | NM_001045516.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010583.mm.1 | TC1100001150.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100010579.mm.1 | NM_001045516.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010579.mm.1 | NM_001045516.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010579.mm.1 | TC1100001150.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100010580.mm.1 | NM_001045516.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010580.mm.1 | NM_001045516.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010580.mm.1 | TC1100001150.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100010585.mm.1 | NM_001045516.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010585.mm.1 | NM_001045516.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010585.mm.1 | TC1100001150.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100010584.mm.1 | NM_001045516.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010584.mm.1 | NM_001045516.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010584.mm.1 | TC1100001150.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100010582.mm.1 | NM_001045516.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010582.mm.1 | NM_001045516.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100010582.mm.1 | TC1100001150.mm.1 | MTA TC |
| Affymetrix HT Mouse Genome 430B Array | 1442508_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1442508_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2534258 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2778555 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2778555 | - | - |

Proca1
Genomic Map