Assay Details
| Gene Symbol: Rnps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080127.1 | NP_001073596.1 | 4 - 5 | 588 | 101 | |
| NM_001080128.1 | NP_001073597.1 | 3 - 4 | 407 | 101 | ||
| NM_009070.2 | NP_033096.2 | 5 - 6 | 710 | 101 | ||
| GenBank mRNA | AK146443.1 | - | 3 - 4 | 406 | 101 | |
| AK151150.1 | - | 4 - 5 | 578 | 101 | ||
| AK165905.1 | - | 3 - 4 | 394 | 101 | ||
| BC002061.1 | - | 4 - 5 | 576 | 3485311 | 101 | |
| BC008089.1 | - | 1 - 2 | 145 | 101 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rnps1
Genomic Map