Assay Details
| Gene Symbol: Akap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035532.2 | NP_001030609.1 | 2 - 3 | 2537 | 82 | |
| NM_001035533.2 | NP_001030610.1 | 1 - 2 | 2478 | 82 | ||
| GenBank mRNA | AF033274.1 | - | 1 - 2 | 2880 | 82 | |
| AF033275.1 | - | 1 - 2 | 2880 | 82 | ||
| AK147420.1 | - | 2 - 3 | 2505 | 82 | ||
| AW489512.1 | - | 1 - 2 | 128 | 82 | ||
| Gene Symbol: Pakap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK077020.1 | - | 3 - 4 | 2942 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Akap2
Genomic Map