| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400035162.mm.1 |
NM_001035532.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400035162.mm.1 |
NM_001035532.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400035162.mm.1 |
NM_001035533.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400035162.mm.1 |
NM_001035533.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400035162.mm.1 |
NM_009649.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400035162.mm.1 |
NM_009649.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400035162.mm.1 |
TC0400004177.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400004524.mm.1 |
NM_001035532.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400004524.mm.1 |
NM_001035532.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400004524.mm.1 |
NM_001035533.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400004524.mm.1 |
NM_001035533.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400004524.mm.1 |
NM_009649.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400004524.mm.1 |
NM_009649.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400004524.mm.1 |
TC0400004177.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400004521.mm.1 |
NM_001035532.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400004521.mm.1 |
NM_001035532.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400004521.mm.1 |
NM_001035533.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400004521.mm.1 |
NM_001035533.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400004521.mm.1 |
NM_009649.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400004521.mm.1 |
NM_009649.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400004521.mm.1 |
TC0400004177.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400004522.mm.1 |
NM_001035532.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400004522.mm.1 |
NM_001035532.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400004522.mm.1 |
NM_001035533.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400004522.mm.1 |
NM_001035533.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400004522.mm.1 |
NM_009649.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400004522.mm.1 |
NM_009649.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400004522.mm.1 |
TC0400004177.mm.1 |
MTA TC |