Assay Details
| Gene Symbol: Rabgef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199059.1 | NP_001185988.1 | 5 - 6 | 684 | 154 | |
| NM_019983.2 | NP_064367.1 | 4 - 5 | 829 | 154 | ||
| GenBank mRNA | AF093590.1 | - | 5 - 6 | 660 | 154 | |
| AK044568.1 | - | 4 - 5 | 849 | 154 | ||
| AK146734.1 | - | 5 - 6 | 634 | 154 | ||
| BC018229.1 | - | 4 - 5 | 829 | 4507477 | 154 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rabgef1
Genomic Map