Assay Details
| Gene Symbol: Sarnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025364.2 | NP_079640.1 | 5 - 6 | 470 | 95 | |
| GenBank mRNA | AK003453.1 | - | 5 - 6 | 322 | 95 | |
| AK018773.1 | - | 5 - 6 | 333 | 95 | ||
| AK088266.1 | - | 5 - 6 | 319 | 95 | ||
| AK146248.1 | - | 5 - 6 | 348 | 95 | ||
| BC027510.1 | - | 5 - 6 | 309 | 1382354 | 95 | |
| BC094393.1 | - | 5 - 6 | 306 | 95 | ||
| BC157909.1 | - | 5 - 6 | 334 | 9088111 | 95 | |
| BC158023.1 | - | 5 - 6 | 334 | 9007329 | 95 | |
| BC172063.1 | - | 5 - 6 | 331 | 9054349 | 95 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sarnp
Genomic Map