Assay Details
| Gene Symbol: Mrpl19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026490.2 | NP_080766.1 | 1 - 2 | 244 | 120 | |
| GenBank mRNA | AK021180.1 | - | 1 - 2 | 238 | 120 | |
| AK030793.1 | - | 1 - 2 | 232 | 120 | ||
| AK050637.1 | - | 1 - 2 | 243 | 120 | ||
| AK141554.1 | - | 1 - 2 | 254 | 120 | ||
| BC020315.2 | - | 1 - 2 | 220 | 120 | ||
| BC043921.1 | - | 1 - 2 | 225 | 5361515 | 120 | |
| BC061064.1 | - | 1 - 2 | 228 | 6535226 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl19
Genomic Map