Assay Details
| Gene Symbol: Gm14920 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102665.1 | NP_001096135.1 | 1 | 270 | 82 | |
| Gene Symbol: H2afb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281531.1 | NP_001268460.1 | 1 | 270 | 82 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2155759 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2062384 | NM_001102665.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2062384 | NM_001102665.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P243045 | NM_001102665.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32617577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32617575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32617561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32617567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32617571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32617554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32617581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32617542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32617578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32617585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32617557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32617576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32617565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32617560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32617583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32617555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32617574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32617584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32617562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32617580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32617568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32617559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32617564 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32617554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32617564 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32617567 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14576786 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32617562 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32617542 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32617557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32617576 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32617561 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32617571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32617575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32617578 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32617560 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32617559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32617580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32617565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32617555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32617572 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32617568 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32617574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32617577 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32617553 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10606553 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601554 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1447894_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10606552 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10601553 | - | - |
| Mouse Genome 430 2.0 Array | 1447894_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1447894_x_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00018383.mm.1 | NM_001281531.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00018383.mm.1 | TC0X00002910.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00007492.mm.1 | NM_001102665.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00007492.mm.1 | NM_001102665.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00007492.mm.1 | TC0X00001163.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1437078_PM_at | - | - |
| Mouse Expression Set 430 | 1437078_at | - | - |
| Mouse Expression Set 430 | 1447894_PM_x_at | - | - |
| Mouse Expression Set 430 | 1447894_x_at | - | - |
| Murine Genome U74v2 Set | 103828_at | - | - |
| Murine Genome U74v2 Set | 103828_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437078_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437078_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447894_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447894_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 437286 | - | - |

Gm14920
Genomic Map