Assay Details
| Gene Symbol: Aadacl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085503.2 | NP_001078972.1 | 1 - 2 | 199 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aadacl3| Gene Symbol: Aadacl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085503.2 | NP_001078972.1 | 1 - 2 | 199 | 81 | |
Set Membership: |
|