Assay Details
| Gene Symbol: Tmem235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085535.1 | NP_001079004.1 | 3 - 4 | 398 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046448 | NM_001085535.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 293319 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 97382 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046448 | NM_001085535.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09619767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167663 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09175689 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09535724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09175694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09535706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167667 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09175724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167595 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09535725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167616 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09195995 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167587 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167592 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167601 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09175685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09619768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09175686 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167619 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09175698 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10383006 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383007 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383008 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10383009 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10383005 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100018839.mm.1 | NM_001085535.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100018839.mm.1 | NM_001085535.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100018839.mm.1 | TC1100001974.mm.1 | MTA TC |

Tmem235
Genomic Map