Assay Details
| Gene Symbol: Prb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198669.1 | NP_941071.1 | 3 | 246 | 134 | |
| GenBank mRNA | BC011176.1 | - | 3 | 246 | 4165240 | 134 |
| Gene Symbol: Gm4736 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053251.1 | NP_444481.1 | 269 | 134 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P417351 | NM_053251.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P417351 | NM_053251.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P71756 | NM_198669.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P71756 | BC011176.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P71756 | NM_198669.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P71756 | BC011176.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969771 | NM_198669.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1969771 | BC011176.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094713 | NM_053251.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 873066 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 131466 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P71756 | NM_198669.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P71756 | BC011176.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P71756 | NM_198669.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P71756 | BC011176.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094713 | NM_053251.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P417351 | NM_053251.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P417351 | NM_053251.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969771 | NM_198669.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1969771 | BC011176.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24852875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24852877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24852863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24852879 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06072205 | - | - |
| Whole Mouse Genome G4122A | A_52_P71756 | NM_198669.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P71756 | BC011176.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P71756 | NM_198669.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P71756 | BC011176.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P417351 | NM_053251.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P417351 | NM_053251.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110354 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121466 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06072205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24852874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24853222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24853022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24852721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24852929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24852766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24852979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24852916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24853090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24852916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24852874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24853121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24852719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24853230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24852834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24852875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24852834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24852830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05289376 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24852874 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24852755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24852877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24852904 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24853144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24852863 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24852825 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24852879 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10548625 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548651 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548645 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548636 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548640 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548644 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548659 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548631 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548642 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548641 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548623 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548652 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548644 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548623 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548624 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548633 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548642 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548657 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548650 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548638 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548628 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548651 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548625 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548628 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548626 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548643 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548641 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548632 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548631 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548634 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548647 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548650 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548633 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548636 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548632 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548626 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548645 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548640 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10548622 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10548656 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10548639 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10548622 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10548639 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600025774.mm.1 | NM_198669.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025774.mm.1 | NM_198669.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025774.mm.1 | TC0600003277.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025776.mm.1 | NM_198669.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025776.mm.1 | NM_198669.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025776.mm.1 | TC0600003277.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025756.mm.1 | TC0600003277.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025777.mm.1 | NM_198669.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025777.mm.1 | NM_198669.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025777.mm.1 | TC0600003277.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025757.mm.1 | TC0600003277.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025779.mm.1 | NM_198669.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025779.mm.1 | NM_198669.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025779.mm.1 | TC0600003277.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025759.mm.1 | XM_003084652.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025759.mm.1 | TC0600003277.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025778.mm.1 | NM_198669.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025778.mm.1 | NM_198669.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025778.mm.1 | TC0600003277.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600025775.mm.1 | NM_198669.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025775.mm.1 | NM_198669.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600025775.mm.1 | TC0600003277.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427745_x_at | - | - |
| Mouse Expression Set 430 | 1427745_PM_x_at | - | - |
| Mouse Expression Set 430 | 1427745_x_at | - | - |
| Mouse Expression Set 430 | 1427745_PM_x_at | - | - |
| Murine Genome U74v2 Set | 93649_f_at | - | - |
| Murine Genome U74v2 Set | 93649_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427745_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427745_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427745_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427745_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 596539 | - | - |
| ABI Mouse Genome Survey Microarray | 695553 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6400446 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006400446 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1257875 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1257875 | - | - |
| mouse-6 v1.1 (Illumina) | scl00114600.1_290-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6400446 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00114600.1_290-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6400446 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3057618 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3134706 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1257875 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3134706 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1257875 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3057618 | - | - |

Prb1
Genomic Map