Assay Details
| Gene Symbol: Krtap11-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113406.1 | NP_001106877.1 | 1 | 547 | 89 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2017347 | NM_001113406.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2006812 | NM_001113406.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2006812 | NM_001113406.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2017347 | NM_001113406.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30951705 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951734 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951758 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951745 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951722 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951724 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951648 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951642 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12572584 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951653 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951714 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12572566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951694 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951740 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951716 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951673 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951667 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951696 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951698 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951671 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951651 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13014920 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12572562 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12853574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951726 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951729 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951703 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951641 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951700 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12607250 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12853580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951719 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951685 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951638 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951666 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12968912 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951735 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951655 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951715 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951660 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951650 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951664 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951759 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951752 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951657 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12572561 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951728 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951661 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951723 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13014918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951736 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951760 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951730 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951658 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951654 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12574144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951713 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951690 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12612737 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12572580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951748 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951646 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30951681 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10440733 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10440732 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439100_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10440731 | - | - |
| Mouse Genome 430 2.0 Array | 1439100_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1439100_s_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600015595.mm.1 | NM_001113406.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600015595.mm.1 | NM_001113406.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600015595.mm.1 | TC1600002037.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1439100_s_at | - | - |
| Mouse Expression Set 430 | 1439100_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439100_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439100_s_at | - | - |

Krtap11-1
Genomic Map