Assay Details
| Gene Symbol: Tsix | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002844.2 | 1 - 2 | 187 | 64 | |||
| GenBank mRNA | AF138745.1 | - | LIT1604 Tsix |
1 - 2 | 187 | 64 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | testis specific X-linked gene | Tsx | AV207184 | -6.9kb | Sense | Chr. X - 103409101 - 103424588 on NCBI Build GRCm38 |
| Downstream | inactive X specific transcripts | Xist |
A430022B11, AI314753 |
12.4kb | Anti-sense | Chr. X - 103460373 - 103483233 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Tsix
Genomic Map