Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger and BTB domain containing 37 | Zbtb37 | D430004I08Rik | -1.1kb | Anti-sense | Chr. 1 - 161008792 - 161034864 on NCBI Build GRCm38 |
| Downstream |
small nucleolar RNA, C/D box 47 |
Snord47 | MBII-96 | 1.5kb | Sense | Chr. 1 - 161038092 - 161038156 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map