Assay Details
| Gene Symbol: Hoxa11os | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015348.1 | 2 - 3 | 787 | 72 | |||
| GenBank mRNA | U20366.1 | - | LIT1565 Hoxa11as |
2 - 3 | 787 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | homeobox A11 | Hoxa11 |
Hox-1.9, Hoxa-11 |
-175.0 | Anti-sense | Chr. 6 - 52242105 - 52245810 on NCBI Build GRCm38 |
| Downstream | homeobox A13 | Hoxa13 |
Hd, Hox-1.10 |
12.3kb | Anti-sense | Chr. 6 - 52258853 - 52260880 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Hoxa11os
Genomic Map