Assay Details
| Gene Symbol: Gt(ROSA)26Sor | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027008.1 | 1 - 2 | 177 | 55 | |||
| GenBank mRNA | U83174.1 | - | LIT3416 Rosa26.1 |
1 - 2 | 130 | 55 | |
| U83175.1 | - | LIT3417 Rosa26.2 |
1 - 2 | 110 | 55 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | THUMP domain containing 3 | Thumpd3 |
AW556087, Gt(ROSA)26asSor, Gtrosa26as |
-2.1kb | Anti-sense | Chr. 6 - 113046252 - 113068278 on NCBI Build GRCm38 |
| Downstream | SET domain containing 5 | Setd5 |
2900045N06Rik, C330007C20, C85544, mKIAA1757 |
427.0 | Anti-sense | Chr. 6 - 113077624 - 113153426 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gt(ROSA)26Sor
Genomic Map