| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600025350.mm.1 |
NM_001098669.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025350.mm.1 |
NM_001098669.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025350.mm.1 |
NM_010653.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025350.mm.1 |
NM_010653.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025350.mm.1 |
TC0600003243.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600025340.mm.1 |
NM_001098669.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025340.mm.1 |
NM_001098669.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025340.mm.1 |
TC0600003243.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600025351.mm.1 |
NM_001098669.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025351.mm.1 |
NM_001098669.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600025351.mm.1 |
TC0600003243.mm.1 |
MTA TC |