| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400015568.mm.1 |
NM_001242605.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015568.mm.1 |
NM_001242605.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015568.mm.1 |
NM_001242606.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015568.mm.1 |
NM_001242606.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015568.mm.1 |
NM_019567.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015568.mm.1 |
NM_019567.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015568.mm.1 |
TC1400002063.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400015569.mm.1 |
NM_001242605.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015569.mm.1 |
NM_001242605.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015569.mm.1 |
NM_001242606.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015569.mm.1 |
NM_001242606.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015569.mm.1 |
NM_019567.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015569.mm.1 |
NM_019567.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015569.mm.1 |
TC1400002063.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400015564.mm.1 |
NM_001085472.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015564.mm.1 |
NM_001085472.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015564.mm.1 |
NM_001085473.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015564.mm.1 |
NM_001085473.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015564.mm.1 |
NM_001242605.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015564.mm.1 |
NM_001242605.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015564.mm.1 |
NM_001242606.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015564.mm.1 |
NM_001242606.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015564.mm.1 |
NM_019567.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015564.mm.1 |
NM_019567.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015564.mm.1 |
NM_023190.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015564.mm.1 |
NM_023190.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400015564.mm.1 |
TC1400002063.mm.1 |
MTA TC |