Assay Details
| Gene Symbol: Abca16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278943.1 | NP_001265872.1 | 26 - 27 | 4462 | 98 | |
| NM_001278944.1 | NP_001265873.1 | 29 - 30 | 4931 | 98 | ||
| GenBank mRNA | AY243472.1 | - | 29 - 30 | 4930 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ATP-binding cassette, sub-family A (ABC1), member 15 |
Abca15 | 4930500I12Rik | -136.3kb | Sense | Chr. 7 - 120328486 - 120407687 on NCBI Build GRCm38 |
| Downstream | Y box protein 1 pseudogene | E130201H02Rik | 52.8kb | Sense | Chr. 7 - 120597625 - 120598475 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2011046 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 93108 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 230653 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 410796 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 737718 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 906479 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 672137 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 543409 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 399040 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 620547 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 291740 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 784956 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 550505 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 656992 | AK148122.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 656992 | BC006796.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 656992 | BC085501.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 656992 | NM_008841.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 656992 | Y13569.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 684489 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 45401 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477612 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441029 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 306514 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 600051 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 891763 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 597942 | AK042798.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 597942 | NM_009399.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 587328 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 152724 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 892618 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 859976 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 347495 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 536637 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 379311 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 315538 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 268656 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 722972 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 952237 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 176952 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 508888 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 426555 | AK031970.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 426555 | AK173294.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 426555 | BC076582.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 426555 | BC079588.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 426555 | BC094219.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 426555 | NM_183141.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 140970 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 773521 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 313542 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 876951 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 624521 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 964928 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 710126 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 940600 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 645106 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 361258 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 183780 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 28596 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 578979 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 360737 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 563797 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 201456 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 848027 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 3513 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 203678 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 900434 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 194214 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477978 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 204021 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 945850 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 193142 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 224333 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 93520 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75100 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 116843 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 131483 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 162490 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 52418 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 111414 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 65291 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 18942 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 11867 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 109170 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 172941 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 178071 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 116728 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 51437 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 102600 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 1074 | - | - |
| Whole Mouse Genome G4122A | A_52_P535430 | AY243472.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P535430 | NR_024051.1 | mouse_transcriptome |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10556902 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556913 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556915 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556926 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556914 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556912 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556925 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556929 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556921 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556916 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556922 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556903 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556931 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556930 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556927 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556905 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556909 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556923 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556908 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556906 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556924 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556919 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556910 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556920 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556904 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556918 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556911 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556928 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556917 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10556907 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1445033_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10556901 | - | - |
| Mouse Genome 430 2.0 Array | 1445033_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1445033_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 233810_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 233810_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700014600.mm.1 | NM_001278943.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700014600.mm.1 | NM_001278944.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700014600.mm.1 | TC0700001727.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1445033_at | - | - |
| Mouse Expression Set 430 | 1445033_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445033_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445033_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 375381 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4560131 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1228982 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1228982 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1228982 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1228982 | - | - |

Abca16
Genomic Map