Assay Details
| Gene Symbol: Abca16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278944.1 | NP_001265873.1 | 7 - 8 | 1110 | 74 | |
| GenBank mRNA | AY243472.1 | - | 7 - 8 | 1110 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ATP-binding cassette, sub-family A (ABC1), member 15 |
Abca15 | 4930500I12Rik | -29.0kb | Sense | Chr. 7 - 120328486 - 120407687 on NCBI Build GRCm38 |
| Downstream | Y box protein 1 pseudogene | E130201H02Rik | 158.3kb | Sense | Chr. 7 - 120597625 - 120598475 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001278944.1 transcripts.
Set Membership: |
|

Abca16
Genomic Map