Assay Details
| Gene Symbol: Asb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003961.1 | 5 - 6 | 997 | 73 | ||
| GenBank mRNA | AK051017.1 | - | 5 - 6 | 910 | 73 | |
| BC062948.1 | - | 1 - 2 | 151 | 30532932 | 73 | |
| BC066824.1 | - | 3 - 4 | 424 | 30087227 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
aldehyde dehydrogenase family 1, subfamily A3 |
Aldh1a3 |
ALDH6, RALDH3, V1 |
-250.7kb | Sense | Chr. 7 - 66390892 - 66427477 on NCBI Build GRCm38 |
| Downstream | lines homolog (Drosophila) | Lins |
2700083B01Rik, AW547219, Lins2, Wins2 |
10.4kb | Anti-sense | Chr. 7 - 66689767 - 66717256 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC062948.1, BC066824.1 transcripts.
Set Membership: |
|

Asb7
Genomic Map