Assay Details
| Gene Symbol: Gm5476 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002868.1 | 1 | 56 | 97 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | keratin 75 | Krt75 |
4732468K03Rik, AA589387, K6hf, Krt2-6hf, Krtcap1 |
-14.5kb | Sense | Chr. 15 - 101563343 - 101573904 on NCBI Build GRCm38 |
| Downstream | type II keratin Kb16P | Gm5477 |
EG432986, Kb16p |
20.3kb | Anti-sense | Chr. 15 - 101608874 - 101610479 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm5476
Genomic Map