Assay Details
| Gene Symbol: Gm8884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026561.1 | 1 | 146 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
neurexophilin and PC-esterase domain family, member 4 |
Nxpe4 |
C130036J11, D930028F11Rik, Fam55d |
-54.3kb | Sense | Chr. 9 - 48162001 - 48400025 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 39334 |
Gm39334 | 4.7kb | Anti-sense | Chr. 9 - 48459419 - 48461173 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm8884
Genomic Map