Assay Details
| Gene Symbol: A930012L18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026853.1 | 4 - 5 | 1474 | 67 | ||
| GenBank mRNA | AK044430.1 | - | 4 - 5 | 1478 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mutated in colorectal cancers | Mcc |
D18Ertd451e, E330037C19 |
Anti-sense | Chr. 18 - 44425060 - 44812182 on NCBI Build GRCm38 | |
| Downstream | YTH domain containing 2 | Ythdc2 |
3010002F02Rik, BC037178 |
151.5kb | Sense | Chr. 18 - 44827744 - 44889720 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

A930012L18Rik
Genomic Map