Assay Details
| Gene Symbol: Llgl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159404.1 | NP_001152876.1 | 12 - 13 | 1678 | 95 | |
| NM_001159405.1 | NP_001152877.1 | 12 - 13 | 1678 | 95 | ||
| NM_008502.2 | NP_032528.1 | 12 - 13 | 1678 | 95 | ||
| GenBank mRNA | AK084386.1 | - | 4 - 5 | 548 | 95 | |
| AK159412.1 | - | 12 - 13 | 1544 | 95 | ||
| BC006017.1 | - | 3 - 4 | 281 | 95 | ||
| BC050913.1 | - | 12 - 13 | 1523 | 6418169 | 95 | |
| BC055399.1 | - | 12 - 13 | 1645 | 6308689 | 95 | |
| D16141.1 | - | 12 - 13 | 1528 | 95 | ||
More Information
Set Membership: |
|

Llgl1
Genomic Map