Assay Details
| Gene Symbol: Ccdc58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159421.1 | NP_001152893.1 | 1 - 2 | 478 | 103 | |
| GenBank mRNA | CJ094333.1 | - | 1 - 2 | 59 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc58
Genomic Map