Assay Details
| Gene Symbol: Ccdc58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159421.1 | NP_001152893.1 | 2 - 3 | 631 | 112 | |
| NM_001159422.1 | NP_001152894.1 | 3 - 4 | 408 | 112 | ||
| NM_198645.2 | NP_941047.1 | 3 - 4 | 757 | 112 | ||
| GenBank mRNA | AK135450.1 | - | 3 - 4 | 337 | 112 | |
| AV583692.1 | - | 3 - 4 | 408 | 112 | ||
| BB686073.1 | - | 1 - 2 | 72 | 112 | ||
| BC024406.1 | - | 3 - 4 | 321 | 112 | ||
| BC024862.1 | - | 3 - 4 | 732 | 5356370 | 112 | |
| BC053690.1 | - | 3 - 4 | 309 | 112 | ||
| CF617453.1 | - | 3 - 4 | 735 | 112 | ||
| CJ094333.1 | - | 2 - 3 | 212 | 112 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ccdc58
Genomic Map