Assay Details
| Gene Symbol: Mafb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010658.3 | NP_034788.1 | 1 | 847 | 76 | |
| GenBank mRNA | AK132425.1 | - | 1 | 848 | 76 | |
| AK143209.1 | - | 1 | 849 | 76 | ||
| AK154830.1 | - | 1 | 209 | 76 | ||
| BC016434.1 | - | 1 | 550 | 4485468 | 76 | |
| BC038256.1 | - | 1 | 795 | 5097650 | 76 | |
| L36435.1 | - | 1 | 627 | 76 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mafb
Genomic Map