Assay Details
| Gene Symbol: Usp44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183199.3 | NP_899022.2 | 3 - 4 | 1004 | 100 | |
| GenBank mRNA | AK088109.1 | - | 3 - 4 | 1005 | 100 | |
| BC111886.1 | - | 1 - 2 | 839 | 40060450 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp44
Genomic Map