Assay Details
| Gene Symbol: 4930534H18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005956.1 | - | 1 | 454 | 103 | |
| AK019718.1 | - | 1 | 511 | 103 | ||
| AV040741.2 | - | 1 | 201 | 103 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1110004E09 gene | 1110004E09Rik | -175.0 | Anti-sense | Chr. 16 - 90925811 - 90934849 on NCBI Build GRCm38 | |
| Downstream | synaptojanin 1 | Synj1 |
A930006D20Rik, AA675315, mKIAA0910 |
524.0 | Anti-sense | Chr. 16 - 90936092 - 91012137 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930534H18Rik
Genomic Map