Assay Details
| Gene Symbol: 1700015H07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005995.1 | - | 1 | 453 | 90 | |
| AK076971.1 | - | 1 | 453 | 90 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 28c | Mir28c | mmu-mir-28c | -7.4kb | Anti-sense | Chr. 15 - 53614205 - 53614279 on NCBI Build GRCm38 |
| Downstream | predicted gene 7489 | Gm7489 | EG665097 | 260.5kb | Sense | Chr. 15 - 53882767 - 53888866 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700015H07Rik
Genomic Map